Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago1/2/3 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
SDP

Catalog No. 501993751
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3751 100 μg
50-199-3752 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3751 Supplier Active Motif Supplier No. 91217
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

The Argonaute family of proteins play roles in RNA interference and in short-interfering-RNA-mediated gene silencing. Small interfering RNAs (siRNAs) have gained much attention for their powerful ability to suppress gene expression. Introduction of double-stranded RNA (dsRNA), that are homologues in sequence to a gene, has proven to suppress that gene's expression through a process known as RNA interference (RNAi); this process post-transcriptionally silences a gene through mRNA inhibition or degradation. Both biochemical and genetic studies have led to the identification of two, conserved gene families that are universal components of the interference process. These are the Dicer family, which is comprised of members of the Ribonuclease III family of enzymes, and the Argonaute gene family. Argonaute proteins addition to their role in RNAi process, may also influence development, such as, stem cell fate determination.

Specifications

Antigen Ago1/2/3
Applications Immunoprecipitation
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5
Gene AGO2
Gene Accession No. Q9H9G7, Q9UKV8, Q9UL18
Gene Symbols AGO1, AGO2, AGO3
Host Species Mouse
Immunogen This Ago1/2/3 antibody was raised against a recombinant protein corresponding to amino acids 47-859 of human Ago2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192669, 26523, 27161
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.