Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGO1/AGO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540075 Supplier Invitrogen™ Supplier No. PA540075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGO1/AGO2 Polyclonal Antibody for Western Blot, ELISA

IEF2C1 and 2 are members of the Argonaute family. These proteins are involved in short-interfering-RNA-mediated gene silencing. This process is mediated by the RISC (RNA induced silencing complex), a multicomponent nuclease that is able to degrade double stranded RNAs. The mRNA degradation is triggered by the association of specific siRNAs with their homologous mRNAs. IEF2C2 has been shown to be essential for embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGO1/AGO2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene AGO2
Gene Accession No. Q9UKV8, Q9UL18
Gene Alias AGO1; AGO2; argonaute 1; argonaute 1, RISC catalytic component; argonaute 2; argonaute 2, RISC catalytic component; argonaute RISC catalytic component 1; Argonaute RISC catalytic component 2; argonaute1; Argonaute2; CTA-204B4.6; EIF2C; eIF2C 1; eIF-2C 1; eIF2C 2; eIF-2C 2; EIF2C1; EIF2C2; Eukaryotic translation initiation factor 2C 1; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C, 1; eukaryotic translation initiation factor 2C, 2; GERP95; Golgi Endoplasmic Reticulum protein 95 kDa; hAgo1; hAgo2; PAZ Piwi domain protein; PPD; protein argonaute-1; Protein argonaute-2; protein slicer; Putative RNA-binding protein Q99; Q10; Q99
Gene Symbols AGO1, AGO2
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide containing a sequence from the C-terminal part of both EIF2C1 and EIF2C2 corresponding to human IEF2C1/2 (eukaryotic translation initiation factor 2C1 and 2).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26523, 27161
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.