Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago1 Monoclonal Antibody (4B8), Active Motif™
SDP

Catalog No. 501993111
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3111 10 μg
50-199-3110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3111 Supplier Active Motif Supplier No. 61072
Add to Cart
Add to Cart

Rat Monoclonal Antibody

AGO1 is a member of the Argonaute family of proteins which play a role in RNA interference. The protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing. AGO1 binds to miRNAs or siRNAs and represses the translation of mRNAs.

Specifications

Antigen Ago1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene Ago1
Gene Accession No. Q9UL18
Gene Alias AGO1; argonaute 1; argonaute 1, RISC catalytic component; Argonaute RISC catalytic component 1; argonaute RISC catalytic subunit 1; Argonaute1; EIF2C; eIF2C 1; eIF-2C 1; Eif2c1; Eukaryotic translation initiation factor 2C 1; eukaryotic translation initiation factor 2C, 1; GERP95; Golgi Endoplasmic Reticulum protein 95 kDa; hAgo1; mAgo1; Piwi/argonaute family protein meIF2C1; Protein argonaute-1; Putative RNA-binding protein Q99; Q99
Gene Symbols Ago1
Host Species Rat
Immunogen This Ago1 antibody was raised against a peptide from the N-terminus of human Argonaute 1 (Ago1) protein.
Purification Method Protein G
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26523
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.