Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ago2/eIF2C2 Antibody (036), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290675MFV610
Description
Ago2/eIF2C2 Monoclonal antibody specifically detects Ago2/eIF2C2 in Mouse samples. It is validated for ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
Ago2/eIF2C2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
036 | |
ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin | |
AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Ago2/eIF2C2 (Uniprot#: Q8CJG0; Met1-Ala860). | |
0.1 mL | |
Epigenetics | |
27161 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction