Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ago2/eIF2C2 Antibody (11A9), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP323228
Description
Ago2/eIF2C2 Monoclonal antibody specifically detects Ago2/eIF2C2 in Human, Bovine samples. It is validated for Western BlotSpecifications
| Ago2/eIF2C2 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Bovine | |
| Purified |
| Western Blot | |
| 11A9 | |
| Western Blot 1:250 | |
| AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10 | |
| Clone 11A9 was generated from an Ago2/eIF2C2 synthetic peptide (coupled to KLH) corresponding to amino acids in the N-terminus of human Ago2/eIF2C2. This sequence has low homology to other argonaute family members. [UniProt# Q9UKV8] | |
| 0.1 mL | |
| Epigenetics | |
| 27161 | |
| Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction