Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ago2/eIF2C2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299647H
Description
Ago2/eIF2C2 Polyclonal antibody specifically detects Ago2/eIF2C2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Ago2/eIF2C2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Ago2/eIF2C2. | |
0.1 mL | |
Epigenetics | |
27161 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
HRP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction