Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago2 Monoclonal Antibody (1B1-E2H5), Active Motif™
SDP

Catalog No. 501992905
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2905 100 μg
50-199-2906 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2905 Supplier Active Motif Supplier No. 39853
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ago proteins are broadly expressed in somatic cells, associate with miRNAs and are key actors in different RNA silencing pathways. However, only the Ago2 protein displays endonucleolytic or 'Slicer' activity and can therefore execute miRNA-directed cleavage of target mRNA, provided that the base-pairing between the Ago2-associated miRNA and the mRNA sequence is perfect. In case of partial complementarity, the Ago2 protein fails to cleave, but instead interferes with translation of the target mRNA via its translational repression activity. Ago2 has been shown to play a non-redundant role in small RNA guided gene silencing processes, including RNA interference, translation repression and heterochromatinization. As a core element of the RISC complex, AGO2 initiates the degradation of target mRNAs through its catalytic activity in gene silencing processes guided by siRNAs or miRNAs. In addition to mRNA degradation or gene silencing guided by miRNAs, Ago2 also acts as a RNA slicer in a Dicer-independent way, as well as a regulator of miRNA maturation. Over-expression of Ago2 has been correlated with several aspects of cancers, including tumor cell growth and the overall survival of cancer patients. Furthermore, gene disruption in the mouse demonstrated that the Ago2 protein is essential for embryonic development and a key regulator of B-lymphoid and erythroid development.

Specifications

Antigen Ago2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1B1-E2H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene AGO2
Gene Accession No. Q9UKV8
Gene Alias 1110029L17Rik; 2310051F07Rik; AGO1; AGO2; AI225898; AL022874; argonaute 2; argonaute 2, RISC catalytic component; Argonaute RISC catalytic component 2; argonaute RISC catalytic subunit 2; Argonaute1; Argonaute2; argonaute2 {ECO:0000255; AW546247; CTA-204B4.6; eIF2C; eIF2C 1; eIF-2C 1; eIF2C 2; eIF-2C 2; eIF2C 2 {ECO:0000255; eIF-2C 2 {ECO:0000255; EIF2C1; Eif2c2; ENSMUSG00000072493; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C 2 {ECO:0000255; eukaryotic translation initiation factor 2C, 2; GERp95; Gm10365; Golgi ER protein 95 kDa; hAgo1; hAgo2; HAMAP-Rule:MF_03031}; Kiaa4215; mAgo2; mKIAA4215; PAZ Piwi domain protein; Piwi/argonaute family protein meIF2C2; PPD; Protein argonaute-1; Protein argonaute-2; Protein slicer; Q10
Gene Symbols AGO2
Host Species Mouse
Immunogen This Ago2 antibody was raised against a recombinant protein corresponding to amino acids 1-148 of human Ago2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27161
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.