Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ago4 Monoclonal Antibody (1B7-G11), Active Motif™

Mouse Monoclonal Antibody
$156.65 - $620.58
Specifications
Antigen | Ago4 |
---|---|
Clone | 1B7-G11 |
Concentration | 1 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2908
![]() ![]() |
Active Motif
39856 |
10 μg |
Each for $156.65
|
|
|||||
50-199-2907
![]() ![]() |
Active Motif
39855 |
100 μg |
Each for $620.58
|
|
|||||
Description
This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic containing PAZ and PIWI domains, and it may play a role in short-interfering-RNA-mediated gene silencing.Specifications
Ago4 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
5730550L01Rik; AGO4; AI481660; argonaute 4; argonaute 4, RISC catalytic component; Argonaute RISC catalytic component 4; argonaute RISC catalytic subunit 4; Argonaute4; eIF2C 4; eIF-2C 4; EIF2C4; Eukaryotic translation initiation factor 2C 4; eukaryotic translation initiation factor 2C, 4; hAgo4; KIAA1567; LOW QUALITY PROTEIN: protein argonaute-4; mAgo4; Piwi/argonaute family protein meIF2C4; Protein argonaute-4 | |
AGO4 | |
IgG | |
Protein G | |
Antibody |
1B7-G11 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9HCK5 | |
192670 | |
This Ago4 antibody was raised against a recombinant protein corresponding to amino acids 1-164 of human Ago4. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
AGO4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title