Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago4 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992875
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2875 100 μL
50-199-2876 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2875 Supplier Active Motif Supplier No. 39813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic containing PAZ and PIWI domains, and it may play a role in short-interfering-RNA-mediated gene silencing.

Specifications

Antigen Ago4
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene AGO4
Gene Accession No. Q9HCK5
Gene Alias 5730550L01Rik; AGO4; AI481660; argonaute 4; argonaute 4, RISC catalytic component; Argonaute RISC catalytic component 4; argonaute RISC catalytic subunit 4; Argonaute4; eIF2C 4; eIF-2C 4; EIF2C4; Eukaryotic translation initiation factor 2C 4; eukaryotic translation initiation factor 2C, 4; hAgo4; KIAA1567; LOW QUALITY PROTEIN: protein argonaute-4; mAgo4; Piwi/argonaute family protein meIF2C4; Protein argonaute-4
Gene Symbols AGO4
Host Species Rabbit
Immunogen This Ago4 antibody was raised against a peptide within the N-terminal region of human Ago4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192670
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.