Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565517 Supplier Invitrogen™ Supplier No. PA565517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQCQVRVLPPV PTEGLTPDDV PALADRVRHS MLTVFREIST DGRGGGDYLK KPG Highest antigen sequence identity to the following orthologs - mouse 89%, rat 93%.

This gene encodes an enzyme that converts lysophosphatidic acid (LPA) into phosphatidic acid (PA). LPA and PA are two phospholipids involved in signal transduction and in lipid biosynthesis in cells. This enzyme localizes to the endoplasmic reticulum. This gene is located in the class III region of the human major histocompatibility complex. Alternative splicing results in two transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AGPAT1
Gene Accession No. Q99943
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 1; 1-acylglycerol-3-phosphate O-acyltransferase 1 (acetoacetly Coenzyme A thiolase); 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, alpha); 1-acyl-sn-glycerol-3-phosphate acyltransferase alpha; 1-AGP; 1-AGP acyltransferase 1; 1-AGPAT; 1-AGPAT 1; 1-AGPAT1; Agpat1; AW047140; G15; LPAATA; LPAAT-alpha; Lysophosphatidic acid acyltransferase alpha; lysophospholipid acyltransferase; Protein G15
Gene Symbols AGPAT1
Host Species Rabbit
Immunogen Recombinant Human AGPAT1. Recombinant protein control fragment (Product #RP-105578).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10554
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.