Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AGPAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103511
Description
Antibody detects endogenous levels of total AGPAT1.
This gene encodes an enzyme that converts lysophosphatidic acid (LPA) into phosphatidic acid (PA). LPA and PA are two phospholipids involved in signal transduction and in lipid biosynthesis in cells. This enzyme localizes to the endoplasmic reticulum. This gene is located in the class III region of the human major histocompatibility complex. Alternative splicing results in two transcript variants encoding the same protein.
Specifications
AGPAT1 | |
Polyclonal | |
Unconjugated | |
AGPAT1 | |
1-acylglycerol-3-phosphate O-acyltransferase 1; 1-acylglycerol-3-phosphate O-acyltransferase 1 (acetoacetly Coenzyme A thiolase); 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, alpha); 1-acyl-sn-glycerol-3-phosphate acyltransferase alpha; 1-AGP; 1-AGP acyltransferase 1; 1-AGPAT; 1-AGPAT 1; 1-AGPAT1; Agpat1; AW047140; G15; LPAATA; LPAAT-alpha; Lysophosphatidic acid acyltransferase alpha; lysophospholipid acyltransferase; Protein G15 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10554, 406165, 55979 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O35083, Q99943 | |
AGPAT1 | |
A synthesized peptide derived from human AGPAT1(Accession Q99943), corresponding to amino acid residues K67-L117. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction