Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AGPAT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598855
Description
AGPAT3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is an acyltransferase that converts lysophosphatidic acid into phosphatidic acid, which is the second step in the de novo phospholipid biosynthetic pathway. The encoded protein may be an integral membrane protein. Two transcript variants encoding the same protein have been found for this gene.
Specifications
AGPAT3 | |
Polyclonal | |
Unconjugated | |
Agpat3 | |
1-acylglycerol-3-phosphate O-acyltransferase 3; 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma; 1-AGP acyltransferase 3; 1-AGPAT 3; Agpat3; AW061257; AW493985; D10Jhu12e; lpaat3; LPAAT-gamma; LPAAT-GAMMA1; lysophosphatidic acid acyltransferase 3; lysophosphatidic acid acyltransferase gamma; lysophosphatidic acid acyltransferase-gamma1; UNQ759/PRO1490 | |
Rabbit | |
Protein G | |
RUO | |
56894 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9NRZ7 | |
Agpat3 | |
Recombinant Human 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma protein (148-304AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction