Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPAT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562768 Supplier Invitrogen™ Supplier No. PA562768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTEIDKGSAY GNSDSKQKLN D Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.

This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AGPAT4
Gene Accession No. Q9NRZ5
Gene Alias 1500003P24Rik; 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase delta); 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, delta); 1-acylglycerol-3-phosphate O-acyltransferase 4; 1-acylglycerol-3-phosphate O-acyltransferase 4 (lysophosphatidic acid acyltransferase, delta); 1-acyl-sn-glycerol-3-phosphate acyltransferase delta; 1-AGP acyltransferase 4; 1-AGPAT 4; 1-AGPAT4; AGPAT4; dJ473J16.2; LPAAT-delta; lysophosphatidic acid acyltransferase delta; lysophosphatidic acid acyltransferase-delta; lysophosphatidic acid acyltransferase-delta (LPAAT-delta); UNQ499/PRO1016
Gene Symbols AGPAT4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AGPAT4. Recombinant protein control fragment (Product #RP-101093).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.