Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGPAT5 Antibody (OTI2C4), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272002AF532
Description
AGPAT5 Monoclonal specifically detects AGPAT5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
AGPAT5 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
AGPAT5 | |
Full length recombinant protein of human AGPAT5 (NP_060831) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2C4 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acidacyltransferase, epsilon), 1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon, 1-AGPAT 5,1-acylglycerol-3-phosphate O-acyltransferase 5, 1AGPAT5,1-AGP acyltransferase 5, EC 2.3.1.51, FLJ11210, LPAAT-e, LPAATE, LPAAT-epsilon, Lysophosphatidic acid acyltransferase epsilon | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
55326.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction