Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPAT5 Monoclonal Antibody (OTI1D12)
GREENER_CHOICE

Catalog No. PIMA525843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525843 Supplier Invitrogen™ Supplier No. MA525843
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AGPAT5 Monoclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT5
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D12
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AGPAT5
Gene Accession No. Q9NUQ2
Gene Alias 1110013A05Rik; 1-acylglycerol-3-phosphate O-acyltransferase 5; 1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acid acyltransferase, epsilon); 1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon; 1-AGP acyltransferase 5; 1-AGPAT 5; 1AGPAT5; 1-AGPAT5; AGPAT5; D8Ertd319e; EC 2.3.1.51; LPAATE; LPAAT-epsilon; lysophosphatidic acid acyltransferase epsilon; RGD1306405; testicular tissue protein Li 144
Gene Symbols AGPAT5
Host Species Mouse
Immunogen Full length human recombinant protein of AGPAT5 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55326
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.