Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPAT5 Monoclonal Antibody (OTI1D4), TrueMAB™, OriGene
SDP

Catalog No. 502540492
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0492 30 μL
50-167-4890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0492 Supplier OriGene Supplier No. TA504140S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AGPAT5 Monoclonal antibody specifically detects AGPAT5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen AGPAT5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KCNJ3
Gene Accession No. P48549
Gene Alias 1110013A05Rik; 1-acylglycerol-3-phosphate O-acyltransferase 5; 1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acid acyltransferase, epsilon); 1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon; 1-AGP acyltransferase 5; 1-AGPAT 5; 1AGPAT5; 1-AGPAT5; AGPAT5; D8Ertd319e; EC 2.3.1.51; LPAATE; LPAAT-epsilon; lysophosphatidic acid acyltransferase epsilon; RGD1306405; testicular tissue protein Li 144
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 279-501 of human KCNJ3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3760
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.