Learn More
AGPAT5 Mouse anti-Human, Clone: OTI1D12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504192

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis.Specifications
AGPAT5 | |
Monoclonal | |
Unconjugated | |
AGPAT5 | |
1AGPAT5, LPAATE | |
Mouse | |
Affinity Chromatography | |
RUO | |
55326 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1D12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NUQ2 | |
AGPAT5 | |
Full length human recombit protein of human AGPAT5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.