Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AGPAT5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532061
Description
Recommended positive controls: A549. Predicted reactivity: Mouse (88%), Rat (87%), Pig (91%), Rhesus Monkey (98%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis.
Specifications
AGPAT5 | |
Polyclonal | |
Unconjugated | |
AGPAT5 | |
1110013A05Rik; 1-acylglycerol-3-phosphate O-acyltransferase 5; 1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acid acyltransferase, epsilon); 1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon; 1-AGP acyltransferase 5; 1-AGPAT 5; 1AGPAT5; 1-AGPAT5; AGPAT5; D8Ertd319e; EC 2.3.1.51; LPAATE; LPAAT-epsilon; lysophosphatidic acid acyltransferase epsilon; RGD1306405; testicular tissue protein Li 144 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55326 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.9 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NUQ2 | |
AGPAT5 | |
Recombinant fragment corresponding to a region within amino acids 45 and 264 of Human AGPAT5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction