Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPAT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552721 Supplier Invitrogen™ Supplier No. PA552721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VAFDCMKNYL DAIYDVTVVY EGKDDGGQRR ESPTMTEFLC KECPKIHIHI DRIDKKDVPE EQEHMRRWLH ERFEIKDKML IEFYESPDPE RRKRFPGKSV NSKLSIKKTL PSMLILSGLT AGMLMTDAGR KLY Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 72%.

This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AGPAT5
Gene Accession No. Q9NUQ2
Gene Alias 1110013A05Rik; 1-acylglycerol-3-phosphate O-acyltransferase 5; 1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acid acyltransferase, epsilon); 1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon; 1-AGP acyltransferase 5; 1-AGPAT 5; 1AGPAT5; 1-AGPAT5; AGPAT5; D8Ertd319e; EC 2.3.1.51; LPAATE; LPAAT-epsilon; lysophosphatidic acid acyltransferase epsilon; RGD1306405; testicular tissue protein Li 144
Gene Symbols AGPAT5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AGPAT5. Recombinant protein control fragment (Product #RP-89849).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55326
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.