Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGPAT5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | AGPAT5 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AGPAT5 Polyclonal antibody specifically detects AGPAT5 in Human samples. It is validated for ImmunofluorescenceSpecifications
AGPAT5 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.2, 40% glycerol | |
55326 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acidacyltransferase, epsilon), 1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon, 1-AGPAT 5,1-acylglycerol-3-phosphate O-acyltransferase 5, 1AGPAT5,1-AGP acyltransferase 5, EC 2.3.1.51, FLJ11210, LPAAT-e, LPAATE, LPAAT-epsilon, Lysophosphatidic acid acyltransferase epsilon | |
This antibody was developed against Recombinant Protein corresponding to amino acids: ANHQSTVDWIVADILAIRQNALGHVRYVLKEGLKWLPLYGCYFAQHGGIYVKRSAKFNEKEMRNKLQSYVDAGTPMYLVIFPEGTRYNPEQTKVLSASQAFAAQRGLAVLKHVLTPRIK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title