Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPAT6 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP348199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP348199 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP348199 Supplier Novus Biologicals Supplier No. NBP348199
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGPAT6 Polyclonal antibody specifically detects AGPAT6 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot

Specifications

Antigen AGPAT6
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:20-1:200, Immunoprecipitation 1:500-1:5000
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acidacyltransferase, zeta), Acyl-CoA:glycerol-3-phosphate acyltransferase 4, DKFZp586M1819,1-acyl-sn-glycerol-3-phosphate acyltransferase zeta, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 4,1-AGP acyltransferase 6, glycerol-3-phosphate acyltransferase 6,1-AGPAT 6, GPAT4,1-acylglycerol-3-phosphate O-acyltransferase 6, LPAAT-zetaLPAATZ, Lysophosphatidic acid acyltransferase zeta, testis spermatogenesis apoptosis-related protein 7, TSARG7
Host Species Rabbit
Immunogen 1-acylglycerol-3-phosphate O-acyltransferase 6(lysophosphatidic acid acyltransferase, zeta). (Uniprot# Q86UL3)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 137964
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.