Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPAT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553428 Supplier Invitrogen™ Supplier No. PA553428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAWATLRMER GAKEKNHQLY KPYTNGIIAK DPTSLEEEIK EIRRSGSSKA LDNTPEFELS DIFYFCRKGM ETIMDDEVTK RFSAEELESW Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPAT4
Gene Accession No. Q86UL3
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 6; 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta); 1-acyl-sn-glycerol-3-phosphate acyltransferase zeta; 1-AGP; 1-AGP acyltransferase 6; 1-AGPAT 6; acyl-CoA:glycerol-3-phosphate acyltransferase 4; AGPAT6; AU041707; AW545732; Glycerol-3-phosphate acyltransferase 4; glycerol-3-phosphate acyltransferase 6; glycerol-3-phosphate acyltransferase-4; Gpat4; LPAATZ; LPAAT-zeta; lysophosphatidic acid acyltransferase zeta; putative lysophosphatidic acid acyltransferase; RGD1310520; testis spermatogenesis apoptosis-related protein 7; TSARG7; UNQ551/PRO1108
Gene Symbols GPAT4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AGPAT6. Recombinant protein control fragment (Product #RP-91086).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137964
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.