Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPAT9 Monoclonal Antibody (OTI1D11)
GREENER_CHOICE

Catalog No. PIMA527208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527208 Supplier Invitrogen™ Supplier No. MA527208
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AGPAT9 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the lysophosphatidic acid acyltransferase protein family. The encoded protein is an enzyme which catalyzes the conversion of glycerol-3-phosphate to lysophosphatidic acid in the synthesis of triacylglycerol. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GPAT3
Gene Accession No. Q53EU6
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-acylglycerol-3-phosphate O-acyltransferase 9; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 10; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 9; 1-AGP acyltransferase 9; 1-AGPAT; 1-AGPAT 9; 4933407I02Rik; 4933408F15; 9; A230097K15Rik; acyl-CoA:glycerol-3-phosphate acyltransferase 3; AGPAT 10; AGPAT10; AGPAT8; AGPAT9; endoplasmic reticulum associated GPAT; glycerol-3-phosphate acyltransferase 3; GPAT3; GPAT-3; hGPAT3; HMFN0839; LPAAT-theta; lung cancer metastasis-associated protein 1; Lysophosphatidic acid acyltransferase theta; lysophosphatidic acid acyltransferase, theta; MAG1; MAG-1; mGPAT3; testis secretory sperm-binding protein Li 213e; UNQ2753/PRO6492
Gene Symbols GPAT3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 41-131 of AGPAT9 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84803
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.