Learn More
AGPAT9 Mouse anti-Human, Clone: OTI3H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809769

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the lysophosphatidic acid acyltransferase protein family. The encoded protein is an enzyme which catalyzes the conversion of glycerol-3-phosphate to lysophosphatidic acid in the synthesis of triacylglycerol. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
AGPAT9 | |
Monoclonal | |
Unconjugated | |
GPAT3 | |
AGPAT 10, AGPAT10, AGPAT8, AGPAT9, HMFN0839, LPAAT-theta, MAG1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84803 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q53EU6 | |
GPAT3 | |
Human recombit protein fragment corresponding to amino acids 41-131 of human AGPAT9 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.