Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPAT9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556137 Supplier Invitrogen™ Supplier No. PA556137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILVKTLEWAT IRIEKGTPKE SILKNSASVG IIQRDESPME KGLSGLRGRD FELSDVFYFS KKGLEAIVED EVTQRFSSEE LVSWNLLTRT N Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.

This gene encodes a member of the lysophosphatidic acid acyltransferase protein family. The encoded protein is an enzyme which catalyzes the conversion of glycerol-3-phosphate to lysophosphatidic acid in the synthesis of triacylglycerol. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPAT3
Gene Accession No. Q53EU6
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-acylglycerol-3-phosphate O-acyltransferase 9; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 10; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 9; 1-AGP acyltransferase 9; 1-AGPAT; 1-AGPAT 9; 4933407I02Rik; 4933408F15; 9; A230097K15Rik; acyl-CoA:glycerol-3-phosphate acyltransferase 3; AGPAT 10; AGPAT10; AGPAT8; AGPAT9; endoplasmic reticulum associated GPAT; glycerol-3-phosphate acyltransferase 3; GPAT3; GPAT-3; hGPAT3; HMFN0839; LPAAT-theta; lung cancer metastasis-associated protein 1; Lysophosphatidic acid acyltransferase theta; lysophosphatidic acid acyltransferase, theta; MAG1; MAG-1; mGPAT3; testis secretory sperm-binding protein Li 213e; UNQ2753/PRO6492
Gene Symbols GPAT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AGPAT9. Recombinant protein control fragment (Product #RP-90973).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84803
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.