Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGPS/Alkyl-DHAP synthase Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12462R
Description
This gene is a member of the FAD-binding oxidoreductase/transferase type 4 family. It encodes a protein that catalyzes the second step of ether lipid biosynthesis in which acyl-dihydroxyacetonephosphate (DHAP) is converted to alkyl-DHAP by the addition of a long chain alcohol and the removal of a long-chain acid anion. The protein is localized to the inner aspect of the peroxisomal membrane and requires FAD as a cofactor. Mutations in this gene have been associated with rhizomelic chondrodysplasia punctata, type 3 and Zellweger syndrome.Specifications
| AGPS/Alkyl-DHAP synthase | |
| Polyclonal | |
| Unconjugated | |
| AGPS | |
| 5832437L22; 9930035G10Rik; AAG5; Adaps; ADAP-S; ADAS; ADHAPS; ADPS; Aging-associated gene 5 protein; aging-associated protein 5; Agps; ALDHPSY; Alkyl-DHAP synthase; Alkyldihydroxyacetonephosphate synthase, peroxisomal; alkylglycerone phosphate synthase; Alkylglycerone-phosphate synthase; AW123847; bs2; RCDP3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8540 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| O00116 | |
| AGPS | |
| KLH conjugated synthetic peptide derived from human AGPS/Alkyl-DHAP synthase. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction