Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGPS Antibody (AGPS-03), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262219MFV610
Description
AGPS Monoclonal antibody specifically detects AGPS in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
AGPS | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
8540 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
AGPS-03 | |
Flow Cytometry, Flow (Intracellular) | |
ADAP-S, ADAS, ADHAPSAlkylglycerone-phosphate synthase, ADPS, Aging-associated gene 5 protein, aging-associated protein 5, ALDHPSY, Alkyl-DHAP synthase, alkyldihydroxyacetonephosphate synthase, peroxisomal, alkylglycerone phosphate synthase, DKFZp762O2215, EC 2.5.1.26, FLJ99755 | |
recombinant human AGPS (amino acids 158-384) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction