Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGPS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587935 Supplier Invitrogen™ Supplier No. PA587935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKIRPVPEYQ KYGSVAFPNF EQGVACLREI AKQRCAPASI RLMDNKQFQF GHALKPQVSS IFTSFLDGLK KFYITKFKGF DPNQLSVATL LFEGDREKVL QHEKQVYDIA AKFGGLAAGE DNGQRGYLLT YVIAYIRDLA LEYYVLGESF ETSAPWDRVV DLCRNVKERI TRECKEKGVQ FAPFSTCRVT QTYDAGACIY FYFAFNYRGI SDPLTVFEQT EAAAREEILA NGGSLSHHHG VGKLRKQWLK ESISDVGFGM LKSVKEYVDP NNIFGNRNLL; Positive Samples: HL-60, HeLa, HepG2, K-562, Mouse kidney, Mouse lung; Cellular Location: Peroxisome membrane.

This gene is a member of the FAD-binding oxidoreductase/transferase type 4 family. It encodes a protein that catalyzes the second step of ether lipid biosynthesis in which acyl-dihydroxyacetonephosphate (DHAP) is converted to alkyl-DHAP by the addition of a long chain alcohol and the removal of a long-chain acid anion. The protein is localized to the inner aspect of the peroxisomal membrane and requires FAD as a cofactor. Mutations in this gene have been associated with rhizomelic chondrodysplasia punctata, type 3 and Zellweger syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPS
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AGPS
Gene Accession No. O00116, Q8C0I1
Gene Alias 5832437L22; 9930035G10Rik; AAG5; Adaps; Adap-s; ADAS; Adhaps; ADPS; Ads; Aging-associated gene 5 protein; aging-associated protein 5; AGPS; ALDHPSY; alkyl-DHAP synthase; alkyldihydroxyacetone phosphate synthase; alkyl-dihydroxyacetonephosphate synthase; alkyldihydroxyacetonephosphate synthase, peroxisomal; alkylglycerone phosphate synthase; alkylglycerone phosphate synthase-like; alkylglycerone-phosphate synthase; AW123847; bs2; RCDP3
Gene Symbols AGPS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 379-658 of human AGPS (NP_0036501).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228061, 8540
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.