Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGR2 Monoclonal Antibody (10E2)
GREENER_CHOICE

Catalog No. PIMA516244
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516244 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516244 Supplier Invitrogen™ Supplier No. MA516244
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sequence homology: Orangutan, Panda and Platypus. Immunogen sequence similarity with other species: Xenopus (82%), Chicken (83%).

AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGR2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10E2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Agr2
Gene Accession No. O88312, O95994
Gene Alias AG2; AG-2; AGR2; agr2 protein; Agr2h; AII; AII type II arginase; anterior gradient 2; anterior gradient 2 homolog; anterior gradient 2 homologue; anterior gradient 2, protein disulphide isomerase family member; anterior gradient homolog 2; anterior gradient protein 2 homolog; ARG2; arginase 2; arginase II; arginase type II; arginase, type II; arginase-2, mitochondrial; AU022422; epididymis secretory protein Li 116; Gob4; Gob-4; HAG-2; HEL-S-116; HPC8; kidney arginase; kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; mAG-2; nonhepatic arginase; Non-hepatic arginase; PDIA17; protein disulfide isomerase family A, member 17; protein Gob-4; secreted cement gland homolog; secreted cement gland protein XAG-2 homolog; Type II arginase; UNQ515/PRO1030; XAG-2
Gene Symbols Agr2
Host Species Mouse
Immunogen A portion of amino acids 50-100 of human AGR2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100053075, 10551, 23795, 415112, 482333
Target Species Human, Mouse, Canine, Monkey, Bovine, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.