Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGR2 Mouse anti-Human, Mouse, Clone: IMG10E2, Abnova™

Catalog No. 89933525 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-525 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-525 Supplier Abnova Corporation Supplier No. MAB12751
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human AGR2.

Specifications

Antigen AGR2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone IMG10E2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human AGR2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (4-6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide).
Gene Alias AG2/GOB-4/HAG-2/XAG-2
Gene Symbols AGR2
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 50-100 of human AGR2.
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10551
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.