Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGR2 Mouse anti-Human, Porcine, Clone: 1A8A8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | AGR2 |
|---|---|
| Clone | 1A8A8 |
| Concentration | 0.93 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7174
![]()
|
Proteintech Group Inc
667681IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).Specifications
| AGR2 | |
| 0.93 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AG 2, AG2, AGR2, GOB 4, HAG 2, HPC8, PDIA17, XAG 2 | |
| AGR2 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 1A8A8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| O95994 | |
| 100514482, 10551 | |
| AGR2 Fusion Protein Ag2919 | |
| Primary | |
| -20°C | |
| AGR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title