Learn More
Invitrogen™ AGR3 Monoclonal Antibody (AGR3.1)

Description
This antibody recognizes the epitope HETTDKNLS within the AGR3 (AG3) protein (19-20 kDa); a secreted cytoplasmic protein which can serve as a marker of carcinogenesis.
Specifications
Specifications
| Antigen | AGR3 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | AGR3.1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | AGR3 |
| Gene Accession No. | Q8TD06 |
| Gene Alias | AG3; AG-3; AGR3; anterior gradient 3; anterior gradient 3 homolog; anterior gradient 3, protein disulphide isomerase family member; anterior gradient homolog 3; anterior gradient protein 3; anterior gradient protein 3 homolog; BCMP11; Breast cancer membrane protein 11; E030025L21Rik; Gm888; H PDIA18; HAG3; hAG-3; PDIA18; Protein disulfide isomerase family A, member 18; RGD1565406; UNQ642/PRO1272 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.