Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AGR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527222
Description
Recommended positive controls: A549, MCF-7. Predicted reactivity: Mouse (92%), Rat (92%), Chicken (86%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AGR3 (Anterior Gradient 3) protein, also known as AG3 (hAG-3, HAG3 in human), or BCMP11, is a secreted cytoplasmic protein which is involved in metastasis induction and p53 tumour supressor inhibition. It may serve as molecular marker and potential therapeutic target for hormone-responsive breast tumours. Its Xenopus homolog is associated with anteroposterior fate determination during early development.
Specifications
| AGR3 | |
| Polyclonal | |
| Unconjugated | |
| AGR3 | |
| AG3; AG-3; AGR3; anterior gradient 3; anterior gradient 3 homolog; anterior gradient 3, protein disulphide isomerase family member; anterior gradient homolog 3; anterior gradient protein 3; anterior gradient protein 3 homolog; BCMP11; Breast cancer membrane protein 11; E030025L21Rik; Gm888; H PDIA18; HAG3; hAG-3; PDIA18; Protein disulfide isomerase family A, member 18; RGD1565406; UNQ642/PRO1272 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 155465, 298959, 403205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8R3W7, Q8TD06 | |
| AGR3 | |
| Recombinant protein encompassing a sequence within the center region of human AGR3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction