Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agrin Mouse anti-Rat, Mouse, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABN120525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1205-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1205-25 Supplier MilliporeSigma Supplier No. MABN120525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Agrin, clone 1H10.1, Cat. No. MABN1205, is a highly specific mouse monoclonal antibody that targets Agrin and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Agrin
Applications Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Agrin Isoform 1
Gene Symbols AGRN;AGRIN
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 99 amino acids from the C-terminal region of human Agrin isoform 1.
Molecular Weight of Antigen ∽220 kDa observed; 217.23 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 1H10.1 detects Agrin in human, mouse, and rat cells. It targets an epitope within 99 amino acids from the C-terminal end.
Target Species Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.