Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AgRP/ART Antibody (04), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP326076MFV450
Description
AgRP/ART Monoclonal antibody specifically detects AgRP/ART in Human samples. It is validated for ELISASpecifications
AgRP/ART | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
04 | |
ELISA | |
agouti related protein homolog (mouse), agouti-related protein, Agrt, AGRTMGC118963, ARTagouti (mouse) related protein, ASIP2 | |
This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
0.1 mL | |
Cardiovascular Biology | |
181 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction