Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AgRP/ART Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | AgRP/ART |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AgRP/ART Polyclonal specifically detects AgRP/ART in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AgRP/ART | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
agouti related protein homolog (mouse), agouti-related protein, Agrt, AGRTMGC118963, ARTagouti (mouse) related protein, ASIP2 | |
AGRP | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Cardiovascular Biology | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
181 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PLKKTTAEQAEEDLLQEAQALAEVLDLQDREPRSSRRCVRLHESCLGQQVPCCDPCATCYCRFFNAFCYCRKLGTAM | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title