Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AgRP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547895
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547895 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547895 Supplier Invitrogen™ Supplier No. PA547895
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 40% cross-reactivity with recombinant mouse AgRP/ART is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Agouti-related protein (AgRP), also called agouti-related peptide, is a neuropeptide produced in the brain by the AgRP/NPY neuron. In humans, the agouti-related peptide is encoded by the AGRP gene. This gene encodes an antagonist of the melanocortin-3 and melanocortin-4 receptor. It appears to regulate hypothalamic control of feeding behavior via melanocortin receptor and/or intracellular calcium regulation, and thus plays a role in weight homeostasis. Mutations in this gene have been associated with late on-set obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AgRP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Agrp
Gene Accession No. O00253
Gene Alias agouti related neuropeptide; agouti related protein; agouti related protein homolog; agouti-related protein; Agrp; AGRT; ART; ASIP2; orexigenic neuropeptide
Gene Symbols Agrp
Host Species Goat
Immunogen Sf 21-derived recombinant human AgRP/ART Ala21-Thr132.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 181
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.