Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ AGS3/GPSM1, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS1531MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1531MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1531MI Supplier MilliporeSigma™ Supplier No. ABS1531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G-protein-signaling modulator 1 (UniProt Q9R080; also known as Activator of G-protein signaling 3) is encoded by the Gpsm1 (also known as Ags3) gene (Gene ID 246254) in rat species. AGS3/GPSM1 is an evolutionarily conserved protein with four G-protein regulatory (GPR) GoLoco motifs (a.a. 493-515, 546-568, 594-616, 628-650 of murine GPSM1) and functions as a guanine nucleotide dissociation inhibitor (GDI). GPSM1 increases heteromeric polycystin-1/polycystin-2 ion channel activity via Gβγ subunits. GPSM1 is involved in regulating mitotic spindle orientation, cell polarity, and adenylyl cyclase activity. GPSM1 plays a role in renal epithelial cell repair following renal injury and GPSM1 induction in polycystic kidney disease (PKD) and may be a critical modulator of the renal cystogenic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGS3/GPSM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene Gpsm1, Ags3
Gene Accession No. Q9R080
Host Species Rabbit
Immunogen Linear peptide corresponding to a sequence within the STK11/LKB1-interacting region of rat AGS3/GPSM1.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_653346
Test Specificity This polyclonal antiserum immunostained predominantly renal collecting ducts of postnatal P12 ADPKD mice. Immunogen peptide blocking eliminated antibody stianing (Kwon, M., et al. (2012). Proc. Natl. Acad. Sci. U. S. A. 109(52):21462-21467). This antiserum targets a STK11/LKB1-interacting region sequence present in rat AGS3/GPSM1 isoform 1 and isoform 4 (isoform Long), 28% present in isoform 3 (Short-2), but completely absent from isoforms 2 (Short-1) reported by UniProt (Q9R080). Target sequence is also present in mouse isoforms 1, 3, 4, and 5, but absent from isoform 2 ( (UniProt Q6IR34).
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.