Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGT Mouse anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980018
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0018 Supplier Bioss Supplier No. BS0045M
Add to Cart
Add to Cart

Mouse Polyclonal Antibody

The protein encoded by AGT, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. Mutations in this gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in this gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.

Specifications

Antigen AGT
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AGT
Gene Accession No. P01019
Gene Alias AGT; AI265500; alpha-1 antiproteinase, antitrypsin; Ang; Ang I; Ang II; Ang III; Ang IV; AngI; AngII; Angiotensin 1-10; Angiotensin 1-4; Angiotensin 1-5; Angiotensin 1-7; Angiotensin 1-8; Angiotensin 1-9; Angiotensin 2-8; Angiotensin 3-8; Angiotensin I; Angiotensin II; Angiotensin III; Angiotensin IV; angiotensin ll; Angiotensin-1; Angiotensin-2; Angiotensin-3; Angiotensin-4; Angiotensinogen; angiotensinogen (PAT); angiotensinogen (serpin peptidase inhibitor, clade A, member 8); ANHU; ANRT; Aogen; Des-Asp[1]-angiotensin II; PAT; pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; Serpin A8; serpin peptidase inhibitor, clade A, member 8; SERPINA8
Gene Symbols AGT
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from rat AGT.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 183
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.