Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGTPBP1 Antibody (OTI9A3), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271734AF647
Description
AGTPBP1 Monoclonal specifically detects AGTPBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
AGTPBP1 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AGTPBP1 | |
Recombinant protein fragment corresponding to amino acids 368-753 of human AGTPBP1 (NP_056054) produced in E.coli. | |
0.1 mL | |
23287.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI9A3 | |
Western Blot, Immunohistochemistry | |
ATP/GTP binding protein 1, ATP/GTP-binding protein 1, CCP1, DKFZp686M20191, EC 3.4.17, EC 3.4.17.-, KIAA1035NNA1cytosolic carboxypeptidase 1, Nervous system nuclear protein induced by axotomy protein 1 homolog, Nna1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction