Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGTPBP1 Monoclonal Antibody (OTI9D11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA800251

Description
NNA1 is a zinc carboxypeptidase that contains nuclear localization signals and an ATP/GTP-binding motif that was initially cloned from regenerating spinal cord neurons of the mouse.Specifications
AGTPBP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UPW5 | |
AGTPBP1 | |
Human recombit protein fragment corresponding to amino acids 368-753 of human AGTPBP1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI9D11 | |
Unconjugated | |
AGTPBP1 | |
CCP1, NNA1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23287 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction