Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGTPBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519400
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519400 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519400 Supplier Invitrogen™ Supplier No. PA519400
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

NNA1 is a zinc carboxypeptidase that contains nuclear localization signals and an ATP/GTP-binding motif that was initially cloned from regenerating spinal cord neurons of the mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGTPBP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene AGTPBP1
Gene Accession No. Q9UPW5
Gene Alias 1700020N17Rik; 2310001G17Rik; 2900054O13Rik; 4930445M19Rik; 5730402G09Rik; agtpbp1; ATP/GTP binding protein 1; ATP/GTP-binding protein 1; carboxypeptidase-tubulin; CCP1; Cytosolic carboxypeptidase 1; cytosolic carboxypeptidase 1; LOW QUALITY PROTEIN: cytosolic carboxypeptidase 1; Cytosolic carboxypeptidase 1-like protein; DKFZp686M20191; KIAA1035; Nervous system nuclear protein induced by axotomy protein 1; nervous system nuclear protein induced by axotomy protein 1 homolog; nmf243; Nna1; nuclear ATP/GTP-binding protein; pcd; Purkinje cell degeneration; si:ch73-236b16.2; soluble carboxypeptidase; tubulinyl-Tyr carboxypeptidase; tyrosine carboxypeptidase
Gene Symbols AGTPBP1
Host Species Goat
Immunogen Synthetic peptide sequence (TSPLEYNLPS) corresponding to the internal amino acids of AGTPBP1 (aa 1141-1151).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23287
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.