Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGTPBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117773 Supplier Invitrogen™ Supplier No. PA5117773
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AGTPBP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

NNA1 is a zinc carboxypeptidase that contains nuclear localization signals and an ATP/GTP-binding motif that was initially cloned from regenerating spinal cord neurons of the mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGTPBP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene AGTPBP1
Gene Accession No. Q9UPW5
Gene Alias 1700020N17Rik; 2310001G17Rik; 2900054O13Rik; 4930445M19Rik; 5730402G09Rik; agtpbp1; ATP/GTP binding protein 1; ATP/GTP-binding protein 1; carboxypeptidase-tubulin; CCP1; Cytosolic carboxypeptidase 1; cytosolic carboxypeptidase 1; LOW QUALITY PROTEIN: cytosolic carboxypeptidase 1; Cytosolic carboxypeptidase 1-like protein; DKFZp686M20191; KIAA1035; Nervous system nuclear protein induced by axotomy protein 1; nervous system nuclear protein induced by axotomy protein 1 homolog; nmf243; Nna1; nuclear ATP/GTP-binding protein; pcd; Purkinje cell degeneration; si:ch73-236b16.2; soluble carboxypeptidase; tubulinyl-Tyr carboxypeptidase; tyrosine carboxypeptidase
Gene Symbols AGTPBP1
Host Species Rabbit
Immunogen E. coli-derived Human AGTPBP1 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23287
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.