Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGTR-1 Antibody (1010103) [Alexa Fluor™ 532], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10244X
Description
AGTR-1 Monoclonal antibody specifically detects AGTR-1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
AGTR-1 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
1010103 | |
Flow Cytometry, CyTOF-ready | |
AG2S, AGTR1AAngiotensin II type-1 receptor, Angiostensin Receptor, angiotensin II receptor, type 1, angiotensin receptor 1, angiotensin receptor 1B, AT1AT1AR, AT1B, AT1R, AT2R1A, AT2R1AGTR1B, AT2R1BAT1BR, HAT1R, type-1 angiotensin II receptor, type-1B angiotensin II receptor | |
Synthetic peptide containing human AGTR-1 | |
0.1 mL | |
GPCR | |
185 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction