Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGTR-2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177368MFV610
Description
AGTR-2 Polyclonal antibody specifically detects AGTR-2 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
AGTR-2 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2ATGR2, MRX88type-2 angiotensin II receptor | |
AGTR2 antibody was raised against a 16 amino acid synthetic peptide from near the center of human AGTR2. The immunogen is located within amino acids 220 - 270 of AGTR2. Amino Acid Squence: LKTNSYGKNRITRDQV | |
0.1 mL | |
Apoptosis, GPCR | |
186 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction