Learn More
Invitrogen™ AGTRAP Polyclonal Antibody

Description
The antibody detects endogenous levels of total AGTRAP protein.
Specifications
Specifications
| Antigen | AGTRAP |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | AGTRAP |
| Gene Accession No. | Q6RW13 |
| Gene Alias | 3300002E14Rik; AGTRAP; angiotensin II receptor associated protein; angiotensin II receptor, type I; angiotensin II receptor-associated protein; angiotensin II, type I receptor-associated protein; AT1 receptor; AT1 receptor-associated protein; AT1R; ATI receptor-associated protein; ATRAP; D4Wsu124e; Type-1 angiotensin II receptor-associated protein |
| Gene Symbols | AGTRAP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.