Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGTRAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560617 Supplier Invitrogen™ Supplier No. PA560617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YHMYRERGGE LLVHTGFLGS SQDRSAYQTI DSAEAPADPF AVPEGRSQDA RGY Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 61%.

This gene encodes a transmembrane protein localized to the plasma membrane and perinuclear vesicular structures. The gene product interacts with the angiotensin II type I receptor and negatively regulates angiotensin II signaling. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGTRAP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AGTRAP
Gene Accession No. Q6RW13
Gene Alias 3300002E14Rik; AGTRAP; angiotensin II receptor associated protein; angiotensin II receptor, type I; angiotensin II receptor-associated protein; angiotensin II, type I receptor-associated protein; AT1 receptor; AT1 receptor-associated protein; AT1R; ATI receptor-associated protein; ATRAP; D4Wsu124e; Type-1 angiotensin II receptor-associated protein
Gene Symbols AGTRAP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AGTRAP. Recombinant protein control fragment (Product #RP-99938).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.