Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AGTRAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576534
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a transmembrane protein localized to the plasma membrane and perinuclear vesicular structures. The gene product interacts with the angiotensin II type I receptor and negatively regulates angiotensin II signaling. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms.
Specifications
AGTRAP | |
Polyclonal | |
Unconjugated | |
AGTRAP | |
3300002E14Rik; AGTRAP; angiotensin II receptor associated protein; angiotensin II receptor, type I; angiotensin II receptor-associated protein; angiotensin II, type I receptor-associated protein; AT1 receptor; AT1 receptor-associated protein; AT1R; ATI receptor-associated protein; ATRAP; D4Wsu124e; Type-1 angiotensin II receptor-associated protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11610, 298646, 57085 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q642A2, Q6RW13, Q9WVK0 | |
AGTRAP | |
Recombinant fusion protein of human AGTRAP. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction