Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGTRAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588400 Supplier Invitrogen™ Supplier No. PA588400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, HT-29, U-87MG, BxPC-3, Mouse kidney, Mouse testis, Mouse heart, Rat kidney, Rat heart; Cellular Location: Cytoplasmic vesicle membrane, Endoplasmic reticulum membrane, Golgi apparatus membrane, Multi-pass membrane protein Immunogen sequence: ILSLLLKPLS CCFVYHMYRE RGGELLVHTG FLGSSQDRSA YQTIDSAEAP ADPFAVPEGR.

This gene encodes a transmembrane protein localized to the plasma membrane and perinuclear vesicular structures. The gene product interacts with the angiotensin II type I receptor and negatively regulates angiotensin II signaling. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGTRAP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AGTRAP
Gene Accession No. Q642A2, Q6RW13, Q9WVK0
Gene Alias 3300002E14Rik; AGTRAP; angiotensin II receptor associated protein; angiotensin II receptor, type I; angiotensin II receptor-associated protein; angiotensin II, type I receptor-associated protein; AT1 receptor; AT1 receptor-associated protein; AT1R; ATI receptor-associated protein; ATRAP; D4Wsu124e; Type-1 angiotensin II receptor-associated protein
Gene Symbols AGTRAP
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11610, 298646, 57085
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.