Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGXT Monoclonal Antibody (1739CT804.82.83.1)
GREENER_CHOICE

Catalog No. PIMA537725
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537725 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537725 Supplier Thermo Scientific Supplier No. MA537725
Add to Cart
Add to Cart

AGXT Monoclonal Antibody (1739CT804.82.83.1)

AGXT Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen AGXT
Applications Western Blot
Classification Monoclonal
Clone 1739CT804.82.83.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene AGXT
Gene Accession No. P21549
Gene Alias AGT; Agt1; Agxt; Agxt1; alanine-glyoxylate aminotransferase; Alanine--glyoxylate aminotransferase; alanine-glyoxylate aminotransferase 1; angiotensin receptor 2; hepatic peroxisomal alanine:glyoxylate aminotransferase; L-alanine: glyoxylate aminotransferase 1; PH1; serine:pyruvate aminotransferase; serine:pyruvate aminotransferase SPT; serine:pyruvate/alanine:glyoxylate aminotransferase; serine-pyruvate aminotransferase; Serine--pyruvate aminotransferase; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, peroxisomal; serine--pyruvate aminotransferase, peroxisomal; serine--pyruvate aminotransferase; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, peroxisomal; Spat; SPT; TLH6
Gene Symbols AGXT
Host Species Mouse
Immunogen recombinant protein between 188-375 amino acids from the human AGXT.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.